missing translation for 'onlineSavingsMsg'
Learn More

Erythrocytes Monoclonal Antibody (SFL23.6), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA135369

429.35 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.01000kg

Artikelnummer. 11520822

  • 644.00 CHF / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

SFL23.6 is directed against an erythroid cell surface antigen, which is not glycophorin A. It shows a well-defined reactivity with cells of the erythroid lineage at all stages of maturation in the peripheral blood, bone marrow, and fetal liver. Non-erythroid lineages are negative by flow cytometry. SFL23.6 is positive on erythroleukemias and can be used to distinguish bone marrow nucleated erythroid precursors from malignant cells in bone marrow specimens

Erythrocytes Monoclonal antibody specifically detects Erythrocytes in Human samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

Erythrocytes
Monoclonal
Unconjugated
Erythroid cell marker; Ly76; Ly-76; TER119
Hepatocytes from 20-22 week-old human fetuses.
100 μg
Primary
4° C
Liquid
Flow Cytometry
SFL23.6
PBS with 0.05% sodium azide
Mouse
purified
RUO
Human
Antibody
IgG2b
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote