missing translation for 'onlineSavingsMsg'
Learn More

Epstein Barr Virus gp250/350 Monoclonal Antibody (C61H), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA17272

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 11500422

  • 594.00 CHF / 100 Mikrogramm
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MA1-7272 antibody reacts with Epstein-Barr virus gp250/350 in viral and infected tissue samples. MA1-7272 has been successfully used in Immunofluorescence and Western blot applications. The MA1-7272 antibody was raised against EBV gp250 protein.

Epstein Barr Virus gp250/350 Monoclonal antibody specifically detects Epstein Barr Virus gp250/350 in Virus samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Epstein Barr Virus gp250/350
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Virus
Antibody
IgG1
Immunocytochemistry, Western Blot
C61H
Unconjugated
EBV gp250; EBV gp350
EBV gp250.
100 μg
Primary
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote