Learn More
Ephrin A3 Monoclonal Antibody (1C12), Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen MA518797
Additional Details : Gewicht : 0.00010kg
Artikelnummer. 15598752
  • 606.00 CHF / 100 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Peptide Sequence: MAAAPLLLLL LLVPVPLLPL LAQGPGGALG NRHAVYWNSS NQHLRREGYT VQVNVNDYLD IYCPHYNSSG VGPGAGPGPG GGAEQYVLYM VSRNGYRTCN ASQGFKRWEC NRPHAPHSPI KFSEKFQRYS AFSLGYEFHA GHEYYYISTP THNLHWKCLR MKVFVCCAST SHSGEKPVPT LPQFTMGPNV KINVLEDFEG ENPQVPKLEK SISGTSPKRE HLPLAVGIAF FLMTFLAS
Ephrin A3 Monoclonal antibody specifically detects Ephrin A3 in Human samples. It is validated for ELISA, Immunoprecipitation.
Spezifikation
Spezifikation
Ephrin A3
Monoclonal
0.34 mg/mL
PBS with no preservative; pH 7.4
P52797
EFNA3
EFNA3 (AAH17722, 1 a.a. ∽ 238 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Immunoprecipitation
1C12
Unconjugated
EFNA3
AW494418; EFL2; EFL-2; EFNA3; EHK1 ligand; EHK1-L; EPH-related receptor tyrosine kinase ligand 3; ephrin A3; ephrin-A3; Epl3; EPLG3; Lerk3; LERK-3; ligand of eph-related kinase 3
Mouse
Affinity chromatography
RUO
1944
-20° C, Avoid Freeze/Thaw Cycles