Learn More
EphA7 Monoclonal Antibody (6C8G7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515457
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-15457 targets EphA7 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15457 immunogen is purified recombinant fragment of EphA7 (aa27-210) expressed in E. Coli.
EphA7 Monoclonal antibody specifically detects EphA7 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
EphA7 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q15375 | |
EPHA7 | |
Purified recombinant fragment of EphA7 (aa27-210) expressed in E. Coli. | |
RUO | |
2045 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
6C8G7 | |
Unconjugated | |
EPHA7 | |
Cek11; Developmental kinase 1; Ebk; EHK3; EHK-3; EK11; embryonic brain kinase; epa7; EPH homology kinase 3; Eph homology kinase-3; EPH receptor A7; Epha7; EPHA7-S; EPH-like kinase 11; ephrin receptor EphA7; ephrin type-A receptor 7; HEK11; Mdk1; mDK-1; receptor protein-tyrosine kinase HEK11; tyrosine-protein kinase receptor EHK-3 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |