Learn More
EphA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529904
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (96%), Pig (97%), Chicken (92%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EphA3 Polyclonal antibody specifically detects EphA3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EphA3 | |
Polyclonal | |
Unconjugated | |
EPHA3 | |
AW492086; Cek4; EK4; End3; EPH receptor A3; EPHA3; EPH-like kinase 4; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; ETK; ETK1; H TYRO4; Hek; Hek4; Human embryo kinase; human embryo kinase 1; I79_002835; Mek4; rEK4; testicular tissue protein Li 64; TYRO4; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor ETK1; tyrosine-protein kinase receptor REK4; tyrosine-protein kinase TYRO4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P29320 | |
EPHA3 | |
Recombinant fragment corresponding to a region within amino acids 38 and 375 of Human Eph Receptor A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |