Learn More
EphA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529315
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2 , HCT116. Predicted reactivity: Mouse (95%), Rat (96%), Chicken (82%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EphA1 Polyclonal antibody specifically detects EphA1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
EphA1 | |
Polyclonal | |
Unconjugated | |
EPHA1 | |
5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; EPH receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; tyrosine-protein kinase receptor EPH; Tyrosine-protein kinase receptor ESK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13835, 2041 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P21709, Q60750 | |
EPHA1 | |
Recombinant fragment corresponding to a region within amino acids 620 and 873 of Human Eph receptor A1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |