missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525358
Additional Details : Gewicht : 0.01000kg
Beschreibung
EpCAM Polyclonal antibody specifically detects EpCAM in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
EpCAM | |
Polyclonal | |
Unconjugated | |
EPCAM | |
adenocarcinoma-associated antigen; CD326; cell surface glycoprotein Trop-1; DIAR5; EGP; EGP-2; EGP314; EGP40; EPCAM; Ep-CAM; EpCAM1; epithelial cell adhesion molecule; Epithelial cell surface antigen; Epithelial glycoprotein; Epithelial glycoprotein 314; ESA; GA733-2; gp40; hEGP314; HNPCC8; human epithelial glycoprotein-2; KS 1/4 antigen; KS1/4; KSA; Ly74; lymphocyte antigen 74; M1S2; M4S1; major gastrointestinal tumor-associated protein GA733-2; mEGP314; membrane component, chromosome 4, surface marker (35kD glycoprotein); MIC18; MK-1; panepithelial glycoprotein 314; protein 289A; Protein D5.7A; Tacsd1; Tacstd1; TROP1; Trop-1 protein; Tumor-associated calcium signal transducer 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P16422 | |
EPCAM | |
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human EpCAM | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |