Learn More
eNOS/NOS Type III Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610296
Additional Details : Gewicht : 0.01000kg
Beschreibung
Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+ levels and enhance calmodulin binding. Neuronal NOS (mNOS) and endothelial NOS (eNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and both are regulated in a similar manner. The human forms exhibit 52% amino acid identity. However, they are distinct gene products of about 155kDa (mNOS) and 140kDa (eNOS). The eNOS gene was cloned from human vascular endothelium as well as from bovine aortic endothelial cells (BAEC). eNOS protein has a unique N-myristylation consensus sequence that may explain its membrane localization.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
eNOS/NOS Type III | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 κ |
Western Blot | |
3 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.