Learn More
ENO3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522275
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2, Molt-4, Raji, mouse brain, Rat2. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (89%), Xenopus laevis (91%), Pig (98%), Rabbit (98%), Chicken (82%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ENO3 Polyclonal antibody specifically detects ENO3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ENO3 | |
Polyclonal | |
Unconjugated | |
ENO3 | |
2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; BBE; beta beta enolase; beta-enolase; cb883; ENO1; ENO3; Eno-3; eno3 protein; enob; enolase 1, (alpha); enolase 3; enolase 3 (beta, muscle); enolase 3, (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; fj24f12; GSD13; hypothetical protein LOC540303; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; Skeletal muscle enolase; wu:fj24f12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13808, 2027, 25438 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P13929, P15429, P21550 | |
ENO3 | |
Recombinant fragment corresponding to a region within amino acids 196 and 434 of ENO3 (Uniprot ID#P13929) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |