missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENO1, Human, Matched Antibody Pair (AP51), Abnova™
To detect and quantify protein level of human
Marke: Abnova H00002023-AP51.1Set
Additional Details : Gewicht : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-ENO1, IgG1 Kappa (100μg) , Detection antibody: rabbit purified polyclonal anti-ENO1 (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
ENO1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ENO1. | |
ENO1L1/MBP-1/MPB1/NNE/PPH | |
Mouse, Rabbit | |
RUO | |
2023 | |
Human | |
Liquid |
ELISA | |
Unconjugated | |
ENO1 | |
ENO1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ENO1, IgG1 Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-ENO1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only