Learn More
ENC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540837
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MSVSVHENRK SRASSGSINI YLFHKSSYAD SVLTHLNLLR QQRLFTDVLL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
ENC1 Polyclonal antibody specifically detects ENC1 in Human samples. It is validated for Western Blot.Spezifikation
ENC1 | |
Polyclonal | |
Unconjugated | |
ENC1 | |
CCL28; ectodermal-neural cortex (with BTB-like domain); ectodermal-neural cortex 1; ectodermal-neural cortex 1 (with BTB domain); ectodermal-neural cortex 1 (with BTB-like domain); Ectoderm-neural cortex protein 1; Enc1; ENC-1; FLJ39259; kelch-like 35; kelch-like 37; kelch-like family member 37; kelch-like protein 37; KLHL35; KLHL37; NRPB; Nuclear matrix protein NRP/B; nuclear restrict protein in brain; nuclear restricted protein, BTB domain-like (brain); p53-induced gene 10 protein; PIG10; si:dkey-35i22.1; TP53I10; tumor protein p53 inducible protein 10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8507 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14682 | |
ENC1 | |
Synthetic peptide directed towards the N-terminal of human ENC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |