Learn More
ELF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540353
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VTLDGIPEVM ETQQVQEKYA DSPGASSPEQ PKRKKGRKTK PPRPDSPATT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%
ELF1 Polyclonal antibody specifically detects ELF1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ELF1 | |
Polyclonal | |
Unconjugated | |
ELF1 | |
E74 like ETS transcription factor 1; E74-like factor 1; E74-like factor 1 (ets domain transcription factor); ELF 1; ELF1; Elf-1; ETS-like transcription factor Elf-1-like protein; ETS-related transcription factor Elf-1; ETS-related transcription factor elf-1-like protein; mElf 1; mElf-1; OTTHUMP00000018310; p70; Sts1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1997 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P32519 | |
ELF1 | |
Synthetic peptide directed towards the N-terminal of human ELF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |