missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523144
Additional Details : Gewicht : 0.01000kg
Beschreibung
EphA2 Polyclonal antibody specifically detects EphA2 in Bovine, Canine, Equine, Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EphA2 | |
Polyclonal | |
Unconjugated | |
EPHA2 | |
ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100053644, 1969, 487421, 512798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
P29317 | |
EPHA2 | |
A portion of amino acids 450-500 of human EphA2 | |
100 μg | |
Primary | |
Bovine, Canine, Equine, Human, Non-human Primate | |
Antibody | |
IgG |