missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAPAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532171
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SAPAP2 Polyclonal antibody specifically detects SAPAP2 in Mouse samples. It is validated for Western Blot.Spezifikation
SAPAP2 | |
Polyclonal | |
Unconjugated | |
DLGAP2 | |
6430596N04Rik; Dap2; DAP-2; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; discs, large (Drosophila) homolog-associated protein 2; disks large-associated protein 2; DLG associated protein 2; Dlgap2; postsynaptic density 95; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 2; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 2; SAPAP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
244310 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8BJ42 | |
DLGAP2 | |
Synthetic peptide corresponding to a region within amino acids 19 and 111 of Mouse Dlgap2 | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |