missing translation for 'onlineSavingsMsg'
Learn More

DHX15 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA541261

Additional Details : Gewicht : 0.00010kg

Artikelnummer. 15584732

  • 604.00 CHF / 100 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Peptide sequence: YINIRKALVT GYFMQVAHLE RTGHYLTVKD NQVVQLHPST VLDHKPEWVL Sequence homology: Cow: 100%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

DHX15 Polyclonal antibody specifically detects DHX15 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

DHX15
Polyclonal
Unconjugated
DHX15
ATP-dependent RNA helicase #46; DBP1; DDX15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 15 (RNA helicase A); DEAD/H box-15; DEAH (Asp-Glu-Ala-His) box helicase 15; DEAH (Asp-Glu-Ala-His) box polypeptide 15; DEAH box protein 15; DEAH9; DEAH-box helicase 15; dhx15; HRH2; im:2639158; mDEAH9; Pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; PRP43; PRPF43; PrPp43p; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15; putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX15-like protein; RNA helicase 2; wu:fb38f09; wu:fk62f05
Rabbit
Affinity Chromatography
RUO
1665
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O43143
DHX15
Synthetic peptide directed towards the C-terminal of human DHX15
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote