missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX24 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541286
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ELRHLLSQPL FTESQKTKYP TQSGKPPLLV SAPSKSESAL SCLSKQKKKK Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%
DDX24 Polyclonal antibody specifically detects DDX24 in Human samples. It is validated for Western Blot.Spezifikation
DDX24 | |
Polyclonal | |
Unconjugated | |
Ddx24 | |
1700055J08Rik; 2510027P10Rik; AI649272; ATP-dependent RNA helicase DDX24; DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; DEAD-box helicase 24; LOC553462 protein; S. cerevisiae CHL1-like helicase; si:ch1073-416d2.2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57062 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9GZR7 | |
Ddx24 | |
Synthetic peptide directed towards the middle region of human DDX24 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |