missing translation for 'onlineSavingsMsg'
Learn More

DDT Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA521386

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13248938

  • 592.00 CHF / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Recommended positive controls: 293T, HeLa, Raji, mouse liver, rat liver. Predicted reactivity: Pig (80%), Rhesus Monkey (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

DDT Polyclonal antibody specifically detects DDT in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

DDT
Polyclonal
Unconjugated
DDT
C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II
Rabbit
Antigen affinity chromatography
RUO
13202, 1652, 29318
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O35215, P30046, P80254
DDT
Recombinant fragment corresponding to a region within amino acids 1 and 118 of DDT (Uniprot ID#P30046)
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote