missing translation for 'onlineSavingsMsg'
Learn More

Cytomegalovirus pp65 Monoclonal Antibody (I1010D), FITC, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA17296

Additional Details : Gewicht : 1.24740kg

Artikelnummer. 11530442

  • 664.00 CHF / 100 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The MA1-7296 antibody reacts with Cytomegalovirus pp65 in viral and infected tissue samples. MA1-7296 has been successfully used in Western blot and Immunofluorescence applications. Acetone fixation of the antigen source is recommended prior to staining. The MA1-7296 antibody was raised against purified CMV pp65.

Cytomegalovirus pp65 Monoclonal antibody specifically detects Cytomegalovirus pp65 in Virus samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Cytomegalovirus pp65
Monoclonal
0.1 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Mouse
purified
RUO
Virus
Antibody
IgG1
Immunocytochemistry, Western Blot
I1010D
FITC
CMV; Cytomegalovirus
Purified CMV pp65.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote