Learn More
Cyclin D1 Mouse anti-Human, Unlabeled, Clone: G124-326, BD
Mouse Monoclonal Antibody
381.00 CHF - 510.00 CHF
Spezifikation
Antigen | Cyclin D1 |
---|---|
Klon | G124-326 |
Konzentration | 0.5mg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827608
|
BD Biosciences
554180 |
0.1 mg |
381.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15837608
|
BD Biosciences
554181 |
0.25 mg |
510.00 CHF
1 Stück |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins contain a conserved amino acid sequence motif, the cyclin box, which allows their binding to cdks to form active complexes that regulate the progression of the cell cycle. The synthesis and degradation of cyclins is tightly controlled in a cell cycle specific manner. Several classes of cyclins (A-E) have been described. Cyclins have been placed into functional groups as follows: Group 1 (cyclins A, B, D1, D2, D3, E and F) functions primarily in cell cycle reguation; Group 2 (cyclins C and H) also plays a role in transcriptional regulation; Group 3 (cyclins G1, G2 and I) may play a role distinct from other cyclins. Specific substrates for cyclin-cdk complexes include nuclear lamins, histones, oncogenes (c-src, c-abl, SV40 large-T Ag), tumor suppressor genes (Rb and p53), and others. The D-type cyclins are involved in regulating the passage of mammalian cells through G1. The reduced molecular weights of cyclin D1 is ∽36kda. G124-326 recognizes human cyclin D1 and cross-reacts with the mouse homolog (Cyl1) of human cyclin D1. It does not cross-react with human cyclins D2 and D3. Recombinant full-length human cyclin D1 was used as immunogen.
Host Species: Mouse
Clone: G124-326
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant full-length human cyclin D1
Formula Weight [Chemical]: 36kDa
Flow Cytometry, Immunoprecipitation, Western Blotting
Spezifikation
Cyclin D1 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
G124-326 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
Recombinant full-length human cyclin D1 | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.