Learn More
Creatine Kinase BB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527509
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, H1299, HeLaS3, HepG2, NIH-3T3. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (85%), Pig (98%), Rabbit (96%), Chicken (89%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Creatine Kinase BB Polyclonal antibody specifically detects Creatine Kinase BB in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Creatine Kinase BB | |
Polyclonal | |
Unconjugated | |
CKB | |
Bck; B-CK; brain creatine kinase; Ck3; Ck-3; CKB; Ckbb; Ckbr; CPK-B; creatine kinase B; creatine kinase B chain; creatine kinase brain; creatine kinase brain-type; Creatine kinase B-type; creatine kinase, brain; creatine kinase-B; Creatine phosphokinase B-type; Creatine phosphokinase M-type; epididymis luminal protein 211; epididymis secretory protein Li 29; HEL-211; HEL-S-29 | |
Rabbit | |
Protein A | |
RUO | |
1152, 12709, 24264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2.05 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P07335, P12277, Q04447 | |
CKB | |
Recombinant fragment corresponding to a region within amino acids 16 and 209 of Human Creatine kinase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |