Learn More
COL14A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531047
Additional Details : Gewicht : 0.01000kg
Beschreibung
PA5-31047 targets COL14A1 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-31047 immunogen is recombit fragment corresponding to a region within amino acids 1027 and 1269 of Human COL14A1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Type XIV collagen is a fibril-associated collagen with an interrupted triple helix. It interacts with the fibril surface and regulates fibrillogenesis (Ansorge et al., 2009 [PubMed 19136672]).Spezifikation
COL14A1 | |
Polyclonal | |
Unconjugated | |
Q05707 | |
COL14A1 | |
Recombit fragment corresponding to a region within amino acids 1027 and 1269 of Human COL14A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
COL14A1 | |
UND, collagen alpha-1(XIV) chain, undulin (fibronectin-tenascin-related) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7373 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |