missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claspin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517227
Additional Details : Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
Claspin Polyclonal antibody specifically detects Claspin in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Claspin | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9HAW4 | |
CLSPN | |
Synthetic peptide corresponding to amino acids near the carboxy terminus of human claspin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
CLSPN | |
B130025E01; C85083; claspin; claspin homolog; CLSPN; E130314M08Rik; hClaspin; MGC131612; MGC131613; MGC131615 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63967 | |
-20°C | |
Liquid |