missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ChAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSC00008W
Additional Details : Gewicht : 0.01000kg
Beschreibung
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μL of sterile water
Spezifikation
ChAT | |
Polyclonal | |
CHAT | |
CHAT | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
P28329, Q03059, P32738 | |
CHAT | |
A synthetic peptide from mouse ChAT | |
RUO | |
1103, 12647, 290567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |