missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Mouse anti-Human, Clone: 3B11, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen LFMA0159
Additional Details : Gewicht : 0.99000kg
Beschreibung
A suggested positive control for this product is human plasma.
The protein encoded by this gene is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe (II)transferrin to Fe (III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.Spezifikation
Ceruloplasmin | |
Monoclonal | |
1 mg/mL | |
CP | |
CP-2 | |
Mouse | |
Protein G | |
RUO | |
1356 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
3B11 | |
Unconjugated | |
P00450 | |
CP | |
Protein purified from Human plasma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |