Learn More
anti-CEBPG Matched Antibody Pair, Abnova
Marke: Abnova H00001054-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-CEBPG (100μg); Mouse polyclonal anti-CEBPG (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP α, C/EBP β, C/EBP γ, and C/EBP δ, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein γ may cooperate with Fos to bind PRE-I enhancer elements.
ELISA
Spezifikation
| CEBPG | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CEBPG. | |
| GPE1BP/IG/EBP-1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CEBPG (100 μg) 2. Detection antibody: mouse polyclonal anti-CEBPG (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| CEBPG | |
| CEBPG | |
| Affinity Purified | |
| RUO | |
| 1054 | |
| Human | |
| Serum |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.