missing translation for 'onlineSavingsMsg'
Learn More

CD9 Monoclonal Antibody (MM2/57), FITC, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA516860

376.01 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15256089

  • 564.00 CHF / 100 Mikrogramm
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Predicted to react with mustelid based on sequence homology. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

CD9 Monoclonal antibody specifically detects CD9 in Bovine, Camelid, Canine, Equine, Feline, Ferret, Human, Mink, Porcine, Rabbit, Rhesus Monkey samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD9
Monoclonal
0.1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
P21926, P30932, P40239, Q8WMQ3
CD9
Human platelet membrane
100 μg
Primary
Bovine, Rhesus Monkey, Camelid, Canine, Feline, Ferret, Ferret, Mink, Porcine, Porcine, Rabbit
Antibody
IgG2b
Flow Cytometry
MM2/57
FITC
CD9
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29
Mouse
Protein A
RUO
100059512, 100141395, 100345844, 280746, 397067, 493874, 611695, 712262, 928
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote