Learn More
CD9 Monoclonal Antibody (EM-04), APC, Invitrogen™
Rat Monoclonal Antibody
Marke:  Invitrogen MA110310
Additional Details : Gewicht : 0.01000kg
Artikelnummer. 12362323
  • 476.00 CHF / 100 Mikrogramm
Verfügbar ab: 24-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry.
Spezifikation
Spezifikation
CD9
Monoclonal
0.5 mg/mL
PBS with 15mM sodium azide; pH 7.4
P40240
CD9
Permeabilized murine bone marrow-derived mast cells (BMMC).
100 μg
Primary
Mouse
Antibody
IgG1
Flow Cytometry
EM-04
APC
CD9
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29
Rat
Size-exclusion chromatography
RUO
12527
4° C, store in dark, DO NOT FREEZE!