missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 beta Monoclonal Antibody (3.4.1), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517588
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD8 beta Monoclonal antibody specifically detects CD8 beta in Rat samples. It is validated for Flow Cytometry.Spezifikation
CD8 beta | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P05541 | |
Cd8b | |
Rat/mouse T cell hybrids expressing CD8 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
3.4.1 | |
Biotin | |
Cd8b | |
CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3 | |
Mouse | |
Protein G | |
RUO | |
24931 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |