missing translation for 'onlineSavingsMsg'
Learn More

CD83 Mouse anti-Human, FITC, Clone: HB15E, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560929

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830609

  • 136.00 CHF / 1 Stück


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The HB15e monoclonal antibody specifically binds to a 45 kDa type 1 transmembrane glycoprotein member of the Ig superfamily. CD83 is composed of a single V-type Ig extracellular domain with a C-terminal cytoplasmic tail. Cell surface CD83 is expressed mainly by follicular dendritic cells, circulating dendritic cells, interdigitating dendritic cells in lymphoid tissues, in vitro-generated dendritic cells and thymic dendritic cells. However, its expression is not restricted to dendritic cells. CD83 is also expressed on some germinal center B cells and some lymphoblastoid cell lines. Although its function is not known, it may play a role in cell-cell interaction during antigen presentation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

CD83
HB15E
BL11; HB15; B-cell activation protein
Affinity Purified
Human CD83 transfected COS cells
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.