Learn More
CD8 Monoclonal Antibody (YTS169.4), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517006
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody recognizes a non-polymorphic epitope on the mouse CD8 alpha chain. It blocks MHC I dependent T cell responses in vitro and in vivo, and induces transplantation tolerance in combination with CD4 antibodies. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD8 Monoclonal antibody specifically detects CD8 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Spezifikation
CD8 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01731, P10300 | |
Cd8a, Cd8b1 | |
The details of the immunogen for this antibody are not currently available. | |
1 mg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen) | |
YTS169.4 | |
Unconjugated | |
Cd8b1 | |
BB154331; CD8; CD8 alpha; CD8 alpha chain; CD8 alpha chain precursor; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha polypeptide; CD8 antigen alpha protein; CD8 antigen alpha protein precursor; CD8 antigen alpha-chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8 beta-2; CD8a; CD8A antigen alpha; CD8a molecule; CD8A; T-cell surface glycoprotein; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; Cd8b1; CD8beta; CD8BP; fCD8; LEU2; Leu-2; Leu2 T-lymphocyte antigen; leu-2a; Ly-2; LY3; Ly-3; Ly-35; Ly-B; Ly-C; Lymphocyte antigen 3; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at | |
Rat | |
Protein G | |
RUO | |
12525, 12526 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |