missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8/CD28 Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Marke: Invitrogen MA112167
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD28 (clone B-23, isotype IgG1).
CD8/CD28 Cocktail antibody specifically detects CD8/CD28 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD8/CD28 | |
Cocktail | |
FITC/PE | |
CD28 | |
CD28; CD28 antigen; CD28 molecule; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; LEU2; Leu-2; Leu2 T-lymphocyte antigen; LY3; LYT3; MAL; MGC138290; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell-specific surface glycoprotein CD28; T-lymphocyte differentiation antigen T8/Leu-2; Tp44 | |
Mouse | |
Protein A/G | |
RUO | |
925, 926, 940 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
17D8, B-23, B-23 | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P01732, P10747, P10966 | |
CD28, CD8A, CD8B | |
human peripheral blood T lymphocytes and HPB-ALL T-cell line | |
50 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |