Learn More
CD79b Mouse anti-Human, PE, Clone: CB3-1, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555679
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunoglobulin (Ig) antigen receptors are composed of a non-covalently- associated complex of Ig and two other proteins, Igα and Igβ, which have been designated in the Fifth International Leukocyte Workshop as CD79a and CD79b respectively. CB3-1 reacts with CD79b, which is expressed on surface Ig(sIg)-positive lymphocytes and B-cell lines but only in the cytoplasm of sIg-negative cells including most terminal deoxynucleotidyl transferase (TdT) positive early pre-B and all cytoplasmic µ positive pre-B cell lines. Antibodies to CD79b are helpful in delineating signal transduction pathways activated via antibody receptors during different stages of B-cell differentiation.
Spezifikation
CD79b | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
CB3-1 | |
Igβ | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
For Research Use Only.