missing translation for 'onlineSavingsMsg'
Learn More

CD71 Mouse anti-Human, PE, Clone: M-A712, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561938

Additional Details : Gewicht : 0.02000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15831489

  • 221.00 CHF / 1 Stück
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The M-A712 monoclonal antibody specifically binds to CD71 which is also known as the transferrin receptor (TFR). This type II transmembrane glycoprotein is expressed on cells as a disulfide-linked homodimer comprised of 95 kDa monomers. CD71 is expressed on activated lymphocytes, monocytes, macrophages, erythroid progenitors, brain endothelium, and most proliferating cells. CD71 is not expressed on resting lymphocytes and is upregulated during lymphocyte responses to antigens or mitogens. Through an endocytic pathway, the transferrin receptor mediates cellular iron uptake by binding and internalizing iron that is bound to transferrin. After releasing iron within the low pH endosomal environment, transferrin and its receptor can be recycled to the cell surface.

Spezifikation

Spezifikation

CD71
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
T-CLL and Jurkat Cells
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
M-A712
TFR; TFRC; Trfr; TfR1; Transferrin receptor protein 1
Mouse
Affinity Purified
RUO
Human
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.