missing translation for 'onlineSavingsMsg'
Learn More

CD62L Mouse anti-Human, PE-CF594, Clone: Dreg 56, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 562301

Additional Details : Gewicht : 0.00840kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15831799

  • 560.00 CHF / 1 Stück


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

TheDREG-56 monoclonal antibody specifically binds to CD62L. CD62L is a 76-95 kDa glycoprotein that is also referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. The DREG-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).

Spezifikation

Spezifikation

CD62L
Dreg 56
SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1
Affinity Purified
Supernatant from PMA-activated Human Peripheral Blood Leukocytes
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.