missing translation for 'onlineSavingsMsg'
Learn More

CD56 Mouse anti-Human, V450, Clone: B159, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560361

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15820139

  • 329.00 CHF / 1 Stück
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reacts with the 220/135kDa heavily glycosylated antigen isoforms present on a subpopulation of peripheral blood large granular lymphocytes which demonstrate natural killer activity, but not on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neutral cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: B159
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Spezifikation

Spezifikation

CD56
Monoclonal
Violet Fluor 450 (V450)
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Human NK Cells
30 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
B159
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.