Learn More
CD56 Mouse anti-Human, V450, Clone: B159, BD
Mouse Monoclonal Antibody
329.00 CHF - 790.00 CHF
Spezifikation
Antigen | CD56 |
---|---|
Klon | B159 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Violet Fluor 450 (V450) |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15820139
|
BD Biosciences
560361 |
30 Tests |
329.00 CHF
1 Stück |
Verfügbar ab: 16-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15810139
|
BD Biosciences
560360 |
120 Tests |
790.00 CHF
1 Stück |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Reacts with the 220/135kDa heavily glycosylated antigen isoforms present on a subpopulation of peripheral blood large granular lymphocytes which demonstrate natural killer activity, but not on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neutral cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: B159
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
CD56 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Human | |
Human NK Cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B159 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 |
For Research Use Only.