Learn More
CD54 (ICAM-1) Mouse anti-Human, Unlabeled, Clone: 28, BD
Mouse Monoclonal Antibody
349.00 CHF
Spezifikation
Antigen | CD54 |
---|---|
Klon | 28 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Beschreibung
The migration of T cells through lymph nodes and their interactions with antigen presenting cells (APCs) involve adhesion molecules on the surfaces of the T cell and the cell with which it interacts. These molecules include the selectins, the integrins, members of the immunoglobulin superfamily, and some mucin-like proteins. The integrins mediate adhesion between cells and between cells and the extracellular matrix. LFA-1 (lymphocyte function-associated antigen 1), a member of the β2 integrin family, is expressed on T cells, neutrophils, and macrophages and is thought to be the most important adhesion molecule for lymphocyte activation. It interacts with members of the immunoglobulin superfamily, the intercellular adhesion molecules: ICAM-1, -2, and -3. ICAM-1 and ICAM-2 are expressed on endodthelium and APCs, while ICAM-3 is expressed only on leukocytes. ICAM-1 is most homologous to neural cell adhesion molecule and myelin-associated glycoprotein. It is a 55kDa glycoprotein that is heavily glycosylated to form 90kDa to 115kDa proteins. Its interactions with LFA-1 are necessary for CTL and NK mediated killing, T and B cell responses, and immune cell extravasation.
Host Species: Mouse
Clone: 28
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CD54/ICAM-1 aa. 46-160
Immunofluorescence, Western Blotting
Spezifikation
CD54 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
IgG1 | |
Affinity Purified |
28 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Primary |
For Research Use Only.