Learn More
CD5 Mouse anti-Human, APC, Clone: UCHT2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561003
Additional Details : Gewicht : 0.01000kg
Beschreibung
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
Spezifikation
CD5 | |
Monoclonal | |
APC | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
UCHT2 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human T cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.