missing translation for 'onlineSavingsMsg'
Learn More

CD49d Mouse anti-Human, PE, Clone: 9F10, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 556635

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15818768

  • 377.00 CHF / 1 Stück


Verfügbar ab
27-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β1 (CD29) or β7. The α4β1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The αβ7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α4 chain-containing integrins.

Spezifikation

Spezifikation

CD49d
9F10
Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4
Affinity Purified
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Immunology
Primary
Baboon, Bovine, Canine, Cynomolgus, Equine, Feline, Human, Rhesus, Sheep
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.