missing translation for 'onlineSavingsMsg'
Learn More

ITGA2 Monoclonal Antibody (AK7), PE, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA516572

373.35 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15215859

  • 560.00 CHF / 100 Tests
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

ITGA2 Monoclonal antibody specifically detects ITGA2 in Human, Non-human Primate samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

ITGA2
Monoclonal
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
P17301
Itga2
The details of the immunogen for this antibody are not currently available.
100 Tests
Primary
Human, Non-human Primate
Antibody
IgG1
Flow Cytometry
AK7
PE
Itga2
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2
Mouse
Protein A
RUO
3673
4° C, store in dark, DO NOT FREEZE!
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote