Learn More
ITGA2 Monoclonal Antibody (AK7), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA180244
Additional Details : Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD49b antibody, clone AK7 recognizes the integrin alpha 2 subunit, a approximately 160 kDa glycoprotein that non-covalently associates with the approximately 130 kDa integrin beta 1 subunit to form the VLA-2 complex.
ITGA2 Monoclonal antibody specifically detects ITGA2 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Spezifikation
ITGA2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P17301 | |
Itga2 | |
Proprietary Immunogen. | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry | |
AK7 | |
Biotin | |
Itga2 | |
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
Mouse | |
Protein A | |
RUO | |
3673 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |