missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD47 Monoclonal Antibody (OX-101), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516893
Additional Details : Gewicht : 0.01000kg
Beschreibung
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD47 Monoclonal antibody specifically detects CD47 in Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Spezifikation
CD47 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P97829 | |
CD47 | |
Rat thymocytes | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
OX-101 | |
Unconjugated | |
CD47 | |
9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
Mouse | |
Protein G | |
RUO | |
29364 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |