missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Monoclonal Antibody (IBL-8), PE, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517988
Additional Details : Gewicht : 0.01000kg
Beschreibung
CD45RC Monoclonal antibody specifically detects CD45RC in Mouse samples. It is validated for Flow Cytometry.Spezifikation
CD45RC | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P06800 | |
Ptprc | |
Mouse Spleen Cells | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
IBL-8 | |
PE | |
Ptprc | |
B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Rat | |
Protein G | |
RUO | |
19264 | |
4° C, do not freeze | |
Liquid |