Learn More
CD45RA Mouse anti-Human, V450, Clone: HI100, BD
Mouse Monoclonal Antibody
353.00 CHF - 760.00 CHF
Spezifikation
Antigen | CD45RA |
---|---|
Klon | HI100 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Violet Fluor 450 (V450) |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15830139
|
BD Biosciences
560363 |
30 Tests |
353.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
12800480
|
BD Biosciences
560362 |
120 Tests |
760.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Reacts with the 220kDa isoform of the human leukocyte common antigen found on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. T cells expressing this antigen are naive or virgin T cells. CD45RA antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: HI100
Isotype: IgG2b, κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining
Spezifikation
CD45RA | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Human | |
IgG2b κ | |
Affinity Purified |
HI100 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.