missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R Monoclonal Antibody (YTH80.103), APC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517693
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-17693 targets CD45R in FACS and WB applications and shows reactivity with Human samples. The MA5-17693 immunogen is rat monoclonal antibody raised against human CD45R.
CD45R Monoclonal antibody specifically detects CD45R in Human samples. It is validated for Flow Cytometry.Spezifikation
CD45R | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.02% sodium azide | |
P08575 | |
PTPRC | |
Rat monoclonal antibody raised against human CD45R | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
YTH80.103 | |
APC | |
PTPRC | |
B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
Rat | |
Protein G | |
RUO | |
5788 | |
4° C, do not freeze | |
Liquid |