Learn More
CD45 Monoclonal Antibody (IBL-5/25), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA170096
Additional Details : Gewicht : 0.01000kg
Description
MA1-70096 detects CD45 in mouse samples. MA1-70096 has been successfully used in flow cytometry, immunohistochemistry (frozen sections) and immunoprecipitation procedures. The MA1-70096 immunogen is mouse lymph node cells.
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation.Spécification
CD45 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P06800 | |
PTPRC | |
Mouse lymph node cells. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
IBL-5/25 | |
Unconjugated | |
PTPRC | |
B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T | |
Rat | |
Protein G | |
RUO | |
19264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |