Learn More
CD45.1 Mouse anti-Mouse, FITC, Clone: A20, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 553775
Additional Details : Gewicht : 0.01000kg
Beschreibung
The A20 monoclonal antibody specifically binds to CD45 (Leukocyte Common Antigen) on all leukocytes of mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, DA). This alloantigen was originally named Ly-5.2, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.2 to Ly-5.1 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb A20 has been reported not to react with leukocytes of most other mouse strains, which express the CD45.2 alloantigen. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C,respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The A20 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.1 alloantigen, to antigens and LPS.
Spezifikation
CD45.1 | |
Monoclonal | |
0.5mg/mL | |
Ly-5; Lyt4; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
A20 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
SJL mouse thymocytes and splenocytes | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.