missing translation for 'onlineSavingsMsg'
Learn More

CD43 Rat anti-Mouse, PE, Clone: S7, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 553271

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15827118

  • 520.00 CHF / 1 Stück
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The S7 monoclonal antibody specifically binds to the 115 kDa glycosylated form of CD43 (Ly-48, Leukosialin). CD43 is expressed on IL-7-responsive pro-B cells, plasma cells, peritoneal and splenic CD5+ B cells (B-1 cells), granulocytes, monocytes, macrophages, platelets, natural killer cells, thymocytes, peripheral T cytotoxic/suppressor cells, and most T helper cells, but not resting conventional peripheral B cells. CD43 expression has also been detected on pluripotent hematopoietic stem cells and myeloid, lymphoid, and NK-cell progenitors in the bone marrow. Studies of CD43-deficient mice indicate that CD43 participates in the negative regulation of T-cell activation and adhesion.

Spezifikation

Spezifikation

CD43
Monoclonal
0.2mg/mL
Spn ; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
S7
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Plasmacytoma MOPC-315
0.2 mg
Immunology
Mouse
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.