Learn More
CD43 Rat anti-Mouse, FITC, Clone: S7, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 553270
Additional Details : Gewicht : 0.01000kg
Beschreibung
The S7 monoclonal antibody specifically binds to the 115 kDa glycosylated form of CD43 (Ly-48, Leukosialin). CD43 is expressed on IL-7-responsive pro-B cells, plasma cells, peritoneal and splenic CD5+ B cells (B-1 cells), granulocytes, monocytes, macrophages, platelets, natural killer cells, thymocytes, peripheral T cytotoxic/suppressor cells, and most T helper cells, but not resting conventional peripheral B cells. CD43 expression has also been detected on pluripotent hematopoietic stem cells and myeloid, lymphoid, and NK-cell progenitors in the bone marrow. Studies of CD43-deficient mice indicate that CD43 participates in the negative regulation of T-cell activation and adhesion.
Spezifikation
CD43 | |
Monoclonal | |
0.5mg/mL | |
Spn ; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
S7 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Plasmacytoma MOPC-315 | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.