missing translation for 'onlineSavingsMsg'
Learn More

CD41 Monoclonal Antibody (PM6/248), PE, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA516919

368.01 CHF gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15266139

  • 552.00 CHF / 100 Tests
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

CD41 Monoclonal antibody specifically detects CD41 in Baboon, Human, Mink, Mustelid samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

CD41
Monoclonal
Conc. Not Determined
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
P08514
Itga2b
Human platelet plasma membrane
100 Tests
Primary
Baboon, Human, Mink, Mustelid
Antibody
IgG1
Flow Cytometry
PM6/248
PE
Itga2b
AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93
Mouse
Protein A
RUO
3674
4° C, store in dark, DO NOT FREEZE!
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote